Furnace Shovel, Steel, Webbers Welding
Catalogue # CA11017-664
Fournisseur: Webber's Welding
Spécifications
- Description:Furnace Shovel
- Cat. no.:CA11017-664
- Supplier No.:FA129
Spécifications
À propos de cet article
The furnace shovel is ideal for either catching scrapings when cleaning the furnace or adding bone ash
The handle is constructed of durable 0.6cm steel rod with a 25.4cm wide x 20.3cm deep pan size for maximum shoveling efficiency
Ordering information: Contact your Avantor sales representative for more information.