Scorifier Steel Tongs, Webber's Welding
Catalog # CA11017-660
Supplier: Webber's Welding
Specifications
- Description:Scorifier Tongs
- Length:40"
- Cat. no.:CA11017-660
- Supplier No.:FA118
Specifications
About this item
The scorifier tongs are made of mild steel for dependable, long-lasting use.
The longer length increases user safety, by distancing the user from the oven or furnace.
Ordering information: Contact your Avantor sales representative for more information.