Cell Strainers
Supplier:   Corning
Cell Strainers
Supplier:  Corning
Certificates
Cell Strainers
3 Options Available Below
Product Details & Documents
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
  • Strong nylon mesh for optimal performance in a variety of applications
  • Extended lip on strainer enables aseptic handling with forceps
  • Moulded color-coded polypropylene frame with tab enables easy handling and identification
  • Sterilised by gamma irradiation, non cytotoxic
  • Conveniently accessible in individual packaging
Use these versatile cell strainers to prepare samples for flow cytometry, single cell suspensions of blood cells, specimens for primary cell cultures and immunogens, as well as primary cells from tissues.
Product Family Options
Informations sur le produit
  • Taille de pore
    Color
    Availability
    Price
  • 40 µm
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $208.05
    $208.05
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $208.05
    Supplier Number: 431750
  • 70 µm
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $209.40
    $209.40
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $209.40
    Supplier Number: 431751
  • 100 µm
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $236.96
    $236.96
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $236.96
    Supplier Number: 431752