ContactUsLinkComponent
Allophycocyanin (Crosslinked APC), Columbia Biosciences
undefined
Allophycocyanin (Crosslinked APC), Columbia Biosciences
  • Storage temperature:
    Store at 4° C
  • Application:
    High Throughput Screening, Fluorescence Resonance Energy Transfer Assays, Flow Cytometry and Microscopy
Frequently Bought Together