Sample Vials with Hinged Lid, Dynalon | Avantor
Sample Vials with Hinged Lid, Dynalon
Supplier:   Dynalon
Sample Vials with Hinged Lid, Dynalon
Supplier:  Dynalon
Certificates
Sample Vials with Hinged Lid, Dynalon
Sample Vials with Hinged Lid, Dynalon
2 Options Available Below
Product Details & Documents
General-purpose containers feature push-in friction fit hinged lids.
Lightweight vials have a wall thickness that is approximately 1 mm. LDPE construction withstands temperatures up to 80 °C (176 °F).
Product Family Options
Informations sur le produit
  • Capacité
    Diameter ×H
    Avec bouchon
    Availability
    Price
  • 7.00 ml
    22.8×32.9 mm
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $637.06
    $637.06
    Case of 100
       $637.06
    Supplier Number: 226245-10
  • 35.00 ml
    31×74.5 mm
    Oui
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $594.86
    $594.86
    Case of 100
       $594.86
    Supplier Number: 226245-50
  • 20.00 ml
    24.8×74.5 mm
    Oui
    Discontinued
    Supplier Number: 226245-25