Formvar Film on Hexagonal Grids, Electron Microscopy Sciences
À propos de cet article
A thin film of pure formvar resin coats the shiny side of a Hexagonal grid.
- Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Select from copper, nickel, and gold grids in 100, 200, 300, and 400 mesh sizes
Optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.
36 Options Available Below
Product Details & Documents
A thin film of pure formvar resin coats the shiny side of a Hexagonal grid.
- Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Select from copper, nickel, and gold grids in 100, 200, 300, and 400 mesh sizes
Optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.
Product Family Options
Informations sur le produit
- DésignationMatièreÉpaisseurMeshAvailabilityPrice
- Supplier Number: FF100H-CU-UL
- Supplier Number: FF200H-CU-UL
- Supplier Number: FF300H-CU-UL
- Supplier Number: FF400H-CU-UL
- Hexagonal GridCopper5 - 6 nm200Discontinued-Supplier Number: FF200H-CU-50
- Hexagonal GridCopper5 - 6 nm300Discontinued-Supplier Number: FF300H-CU-25
- Hexagonal GridCopper5 - 6 nm300Discontinued-Supplier Number: FF300H-CU-50
- Supplier Number: FF100H-CU-TH
- Supplier Number: FF200H-CU-TH
- Supplier Number: FF300H-CU-TH
- Supplier Number: FF400H-CU-TH
- Supplier Number: FF100H-CU-ET
- Supplier Number: FF200H-CU-ET
- Supplier Number: FF300H-CU-ET
- Supplier Number: FF400H-CU-ET
- Supplier Number: FF100H-AU-UL
- Supplier Number: FF200H-AU-UL
- Supplier Number: FF300H-AU-UL
- Supplier Number: FF400H-AU-UL
- Hexagonal GridGold5 - 6 nm200Discontinued-Supplier Number: FF200H-AU-50
- Supplier Number: FF100H-AU-TH
- Supplier Number: FF200H-AU-TH
- Supplier Number: FF300H-AU-TH
- Supplier Number: FF400H-AU-TH
- Supplier Number: FF100H-AU-ET
- Supplier Number: FF200H-AU-ET
- Supplier Number: FF300H-AU-ET
- Supplier Number: FF400H-AU-ET
- Supplier Number: FF100H-NI-UL
- Supplier Number: FF200H-NI-UL
- Supplier Number: FF300H-NI-UL
- Supplier Number: FF400H-NI-UL
- Supplier Number: FF100H-NI-TH
- Supplier Number: FF200H-NI-TH
- Supplier Number: FF300H-NI-TH
- Supplier Number: FF400H-NI-TH
- Supplier Number: FF100H-NI-ET
- Supplier Number: FF200H-NI-ET
- Supplier Number: FF300H-NI-ET
- Supplier Number: FF400H-NI-ET