Sklar Retractors are stainless steel, industry standard.
The Senn Retractor is a handheld, double-ended retractor used to retract primarily surface tissue. It is often used in plastic surgery, small bone and joint procedures, or thyroidectomy and dissection of neck tissue. This instrument has one angled, blunt end and, at the opposite end, facing the opposite direction has a three prong, rake tip. The prongs may be sharp or blunt. They have a straight, flat handle with one squared off blunt end and a second rake end with blunt tips.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 82030-280
- 82030-282
- Disposable / reusable:
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free