API Precision Combined Form Hydrometer, Thermco
API Precision Combined Form Hydrometer, Thermco
Supplier:   Thermco
API Precision Combined Form Hydrometer, Thermco
Supplier:  Thermco
Certificates
API Precision Combined Form Hydrometer, Thermco
API Precision Combined Form Hydrometer, Thermco
8 Options Available Below
Product Details & Documents
Precison API combined form hydrometers made to meet API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and mounted in the body of the hydrometer. Length of the hydrometer is 15 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Product Family Options
Informations sur le produit
  • Plage de température
    Longueur
    Availability
    Price
  • RANGE 10/45 X 0.5
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $145.23
    $145.23
    Each
       $145.23
    Supplier Number: GW6000S
  • RANGE 0/20 X 0.2
    15"
    Discontinued
    Supplier Number: -
  • RANGE 45/90X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.30
    $134.30
    Each
       $134.30
    Supplier Number: GW6001S
  • RANGE 30/90 X 0.5
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.07
    $136.07
    Each
       $136.07
    Supplier Number: GW6001XS
  • RANGE 10/30 X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.30
    $134.30
    Each
       $134.30
    Supplier Number: GW6002S
  • RANGE 20/40 X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.30
    $134.30
    Each
       $134.30
    Supplier Number: GW6003S
  • RANGE 30/50 X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.30
    $134.30
    Each
       $134.30
    Supplier Number: GW6004S
  • RANGE 40/60 X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $136.07
    $136.07
    Each
       $136.07
    Supplier Number: GW6005S
  • RANGE 50/70 X 0.2
    15"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $134.30
    $134.30
    Each
       $134.30
    Supplier Number: GW6006S
  • RANGE 60/80 X 0.2
    15"
    Discontinued
    Supplier Number: -
  • RANGE 40/70 X 0.5
    15"
    Discontinued
    Supplier Number: -
  • RANGE 70/90 X 0.2
    15"
    Discontinued
    Supplier Number: -
  • RANGE 80/100 X 0.2
    15"
    Discontinued
    Supplier Number: -