API Precision Short Plain Form Hydrometer, Thermco
À propos de cet article
Precison API plain form hydrometers made to meet API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers.
Length of the hydrometer is 6 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
10 Options Available Below
Product Details & Documents
Precison API plain form hydrometers made to meet API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometers will perform with the same accuracy as the ASTM hydrometers.
Length of the hydrometer is 6 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: GW2500
- Supplier Number: GW2501
- Supplier Number: GW2502
- Supplier Number: GW2503
- Supplier Number: GW2504
- Supplier Number: GW2505
- Supplier Number: GW2506
- Supplier Number: GW2507
- Supplier Number: GW2508
- Supplier Number: GW2509