ASTM/API Precision Plain Form Hydrometer, Thermco | Avantor
ASTM/API Precision Plain Form Hydrometer, Thermco
Supplier:   Thermco
ASTM/API Precision Plain Form Hydrometer, Thermco
Supplier:  Thermco
Certificates
ASTM/API Precision Plain Form Hydrometer, Thermco
ASTM/API Precision Plain Form Hydrometer, Thermco
26 Options Available Below
Product Details & Documents
Precison ASTM plain form hydrometers made to meet ASTM and API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degrees.
Tall form, 330mm, have 12 degree ranges; Short form, 163mm, have 6 degree ranges.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Product Family Options
Product Information
  • Modèle
    Range
    Longueur
    Subdivisions
    Availability
    Price
  • 3H
    19...31
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $79.77
    $79.77
    Each
       $79.77
    Supplier Number: GW003H
  • 4H
    29...41
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $67.81
    $67.81
    Each
       $67.81
    Supplier Number: GW004H
  • 5H
    39...51
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW005H
  • 6H
    49...61
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $67.81
    $67.81
    Each
       $67.81
    Supplier Number: GW006H
  • 7H
    59...71
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW007H
  • 8H
    69...81
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW008H
  • 9H
    79...91
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW009H
  • 10H
    89...101
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW010H
  • 21H
    0...6
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW021H
  • 22H
    5...11
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW022H
  • 23H
    10...16
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW023H
  • 23H
    15...21
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW024H
  • 25H
    20...36
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW025H
  • 26H
    25...31
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW026H
  • 28H
    35...41
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW028H
  • 29H
    40...46
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW029H
  • 30H
    45...51
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW030H
  • 31H
    50...56
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW031H
  • 1H
    1...11
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW001H
  • 2H
    9...21
    330 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $67.81
    $67.81
    Each
       $67.81
    Supplier Number: GW002H
  • 32H
    55...61
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW032H
  • 33H
    60...66
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW033H
  • 34H
    65...71
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW034H
  • 35H
    70...76
    163 mm
    0.1
    Discontinued
    Supplier Number: -
  • 36H
    75...81
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW036H
  • 37H
    80...86
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW037H
  • 38H
    85...91
    163 mm
    0.1
    Discontinued
    Supplier Number: -
  • 39H
    90...96
    163 mm
    0.1
    Discontinued
    Supplier Number: -
  • 40H
    95...101
    163 mm
    0.1
    Log in for availability
    Log in for availability
    Each
    Each - $66.93
    $66.93
    Each
       $66.93
    Supplier Number: GW040H