Orion™ Basic Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
À propos de cet article
Perform basic pH measurements quickly and reliably with Orion™ Basic Electrodes, for use with Orion™ Russell meters.
- pH range of 0 to 14
3 Options Available Below
Product Details & Documents
Perform basic pH measurements quickly and reliably with Orion™ Basic Electrodes, for use with Orion™ Russell meters.
- pH range of 0 to 14
Specifications for Product Family
Spécifications
- Catalog No:
- 14004-290
- 14004-288
- 14004-294
- Operating temperature range:
- 0 to 80°C
- 0 to 80°C
- 0 to 80°C
Spécifications
Product Family Options
Informations sur le produit
- DésignationMatièreAvailabilityPrice
- Low Maintenance Gel Combination Electrode, with built-in ATCEpoxy BodyLog in pour la disponibilitéEach$456.00Supplier Number: 9147BNSpécifications:
More
Specifications
Cat. No.14004-290Operating temperature range0 to 80°C - Low Maintenance Gel Combination Electrode, with Removable Bulb GuardEpoxy BodyLog in pour la disponibilitéEach$236.29Supplier Number: 9146BNSpécifications:
More
Specifications
Cat. No.14004-288Operating temperature range0 to 80°C - Supplier Number: 9142BNSpécifications:
More
Specifications
Cat. No.14004-294Operating temperature range0 to 80°C