Thymus Lysate, ProSci
Thymus Lysate, ProSci
Supplier:   Prosci
Thymus Lysate, ProSci
Supplier:  Prosci
Certificates
3 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Thymus tissue lysate (Human) was prepared by homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)
Thymus tissue lysate (Mouse and Rat) was prepared by homogenization in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7.4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin. Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 10062-224
  • 10062-268
  • 10062-332
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • 2 mg/mL
  • Tested applications:
  • WB
  • WB
  • WB
  • Storage buffer:
  • Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Storage conditions:
  • Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
Specifications
Product Family Options
Product Information
  • Désignation
    Format de cellule
    Taille
    Source
    Tissue
    Availability
    Price
  • Normal
    Cryopreserved
    0.1 mg
    Mouse
    Thymus
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $341.68
    $341.68
    Each (100µG)
       $341.68
    Minimum order quantity is 1
    Supplier Number: 1409
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10062-224
    Concentration
    2 mg/mL
    Tested applications
    WB
    Storage buffer
    Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
    Storage conditions
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Normal
    Cryopreserved
    0.1 mg
    Rat
    Thymus
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $341.68
    $341.68
    Each (100µG)
       $341.68
    Minimum order quantity is 1
    Supplier Number: 1471
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10062-268
    Concentration
    2 mg/mL
    Tested applications
    WB
    Storage buffer
    Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
    Storage conditions
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Normal
    Cryopreserved
    0.1 mg
    Human
    Thymus
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $332.68
    $332.68
    Each (100µG)
       $332.68
    Minimum order quantity is 1
    Supplier Number: 1314
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10062-332
    Concentration
    2 mg/mL
    Tested applications
    WB
    Storage buffer
    Thymus lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
    Storage conditions
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.