Floor Pad Holders, Remco Products
This pad holder is an excellent tool for cleaning walls and tanks
It is also great for scrubbing stubborn dirt and cleaning underneath equipment. The clip mechanism is stainless steel, which prevents the pad from falling off once heavy with water. Polypropylene material.
: FDA compliant raw material.
: This floor model requires a European-thread handle.
Frequently Bought Together