NOVA-COT ™ Natural Cotton Cleanroom Wipes, High-Tech Conversions
NOVA-COT™ 100% cotton cleanroom wipes are ideal for use in a variety of controlled environments including ISO Class 6 (formerly class 1000) cleanrooms.
- Contains no contaminating additives and is virtually chemically pure
- It is bias cut; therefore, it is very low in particulates and stray fibers
- Virgin cotton is naturally absorbent, non-abrasive, biodegradable, and low in static
NOVA-COT™ cotton cleanroom wipes are woven from the highest grade cotton into a 2×1 twill construction. It is bleached, singed, and mercerized to remove virtually all impurities that are required to meet critical federal government and major semiconductor manufacturer specifications.
: Wipes are packaged 300 wipes per bag, with eight bags per case.
- Catalog No:
- 10152-446
- Basis weight:
- 162.7 gsm
Frequently Bought Together