Pleated (Fluted) Filter Paper Grade 562, Ahlstrom
: Ahlstrom
Ahlstrom Grade 562 is the pre-pleated version of Grade 962.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
- Catalog No:
- 47745-862
- 47745-858
- 47745-860
- 47745-856
- 47745-852
- 47745-854
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- High
- High
- High
- High
- High
- High
- Retention:
- 27 µm
- 27 µm
- 27 µm
- 27 µm
- 27 µm
- 27 µm
Frequently Bought Together