Pleated (Fluted) Filter Paper, Grade 561, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 561 is the pre-pleated version of Grade 961.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Specifications
- Catalog No:
- 47749-644
- 47749-646
- 47749-642
- 47749-650
- 47749-648
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- High
- High
- High
- High
- High
- Retention:
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm