Pleated (Fluted) Filter Paper, Grade 515, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 515 is the pre-pleated version of Grade 615.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Specifications
- Catalog No:
- 47745-848
- 38000-200
- 47745-846
- Flow rate:
- Fast
- Fast
- Fast
- Load capacity:
- High
- High
- High
- Retention:
- 25 µm
- 25 µm
- 25 µm