Pleated (Fluted) Filter Paper, Grade 513, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 513 is the pre-pleated version of Grade 613.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Specifications
- Catalog No:
- 47745-840
- 47745-842
- 71440-004
- 47745-844
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Retention:
- 6.0 µm
- 6.0 µm
- 6.0 µm
- 6.0 µm