Pleated (Fluted) Filter Paper, Grade 542, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Ahlstrom Grade 542 is the pre-pleated version of Grade 642.
Pleating improves the effective surface area which increases flow rate and loading capacity, thus decreasing filtration time.
Specifications
- Catalog No:
- 38002-066
- 38002-044
- 47749-636
- 47749-638
- 47749-634
- 38002-088
- Flow rate:
- Slow
- Slow
- Slow
- Slow
- Slow
- Slow
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm