Hardened Ashless Quantitative Paper, Grade 75, Ahlstrom
: Ahlstrom
Ahlstrom Grade 75 is a medium flow rate hardened ashless (Ash 0.006%) filter paper suitable for quantitative gravimetric analysis in analytical techniques requiring increased chemical resistance, wet strength and handling capacity.
- Catalog No:
- 47745-750
- 47749-494
- 47749-496
- 47745-752
- 47749-490
- 47745-746
- 47745-748
- 47749-492
- 47749-488
- 47749-484
- 47749-486
- 47749-480
- 47749-482
- 47749-478
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
- 2.0 µm
Frequently Bought Together