Wet-Strengthened Qualitative Filter Papers, Grade 962
Fournisseur: Ahlstrom
Certificats
À propos de cet article
Ahlstrom Grade 962 is a fast filtering grade with a creped surface and high loading capacity.
It is a thinner version of Grade 961.
Spécifications
- Catalog No:
- 47749-802
- 47749-804
- 47749-798
- 47749-800
- 47749-794
- 47749-796
- 47749-790
- 47749-812
- 47749-792
- 47749-814
- 47749-806
- 47749-808
- 47749-810
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- High
- Retention:
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm
- 28 µm