Hardened Ashless Quantitative Paper, Grade 55, Ahlstrom
: Ahlstrom
Ahlstrom Grade 55 is a fast filtering hardened ashless (Ash 0.006%) filter paper suitable for quantitative gravimetric analysis of coarse particles and gelatinous precipitates in analytical techniques requiring increased wet strength and handling capacity.
- Catalog No:
- 47749-450
- 47749-452
- 47749-448
- 38008-129
- 47745-738
- 38008-027
- 47749-462
- 47749-458
- 47749-460
- 47749-454
- 47749-456
- 47745-736
- 28334-145
- 28334-154
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Medium
- Retention:
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
Frequently Bought Together