Vikan® Tube Brush, 0.4", Remco Products
This tube brush is the smallest diameter of the tube brushes.
- Dishwasher safe
- Autoclavable up to 250 °F
Block material is polypropylene; bristle material is polyester.
It is used to clean small pipes and very narrow spaces between machine parts. The bristles are attached to a helically wound stainless steel shaft, making it strong and chemically resistant.
: FDA compliant raw material.
Frequently Bought Together