Vikan® Tube Brush, 2.5", Remco Products
Supplier: Remco Products
Certificates
About this item
This tube brush is the largest diameter of the tube brushes. It is used to clean pipes, tab outlets, and narrow spaces between machine parts.
- Dishwasher safe
- Autoclavable up to 250 °F
Block material is polypropylene; Bristle material is polyester.
The bristles are attached to a helically wound stainless steel shaft which makes it chemical resistant and strong.
Certifications: FDA compliant raw material.
Frequently Bought Together