Virtua™ Series Reader Protective Eyewear, 3M™
89095-244
: 3M
- Frame Color:Clear
- Description:Frameless glasses
- Cat. no.:89095-244
- Lens Coating:Anti-fog
- Lens Tint:Clear
- Diopter Strength:1.5
- Supplier No.:11513-00000-20
Virtua™ series reader protective eyewear is designed for workers who have difficulty reading small print or are engaged in detail work.
- Molded-in magnifying diopters
- Unisex style
- Wraparound coverage
- Polycarbonate lens absorbs 99.9% UV
Virtua™ series reader protective eyewear line merges outstanding unisex style, protection, and affordability, all while offering wearers an open and unobstructed viewing range. Virtua™ series reader protective eyewear is a valuable choice in vision safety.
Unisex Virtua™ series reader protective eyewear features magnifying diopters, lightweight lenses with contoured tip temples, and a high wrap for additional coverage.
: Meets Standards ANSI Z87.1-2003