Cavity Well Microscope Slides, Globe Scientific
Fournisseur: Globe Scientific
Certificats
À propos de cet article
Cavity well microscope slides are ideal for holding liquids and cultures for microscopic examination
- Pre-cleaned and ready to use
- Spherically polished cavities are 13 mm diameter with 0.2 to 0.4 mm depth
- Dimensions: 25×75×1 mm
All slides are pre-cleaned and supplied ready for use. The spherically polished cavities are 13 mm.
They feature 90° ground edges with slightly clipped corners for safety. Two styles are available, with a single or double cavity.
Packaging: 72/box, 20 boxes/case (10 gross)
Frequently Bought Together