Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-EIF2AK2 Rabbit Polyclonal Antibody | Avantor

Anti-EIF2AK2 Rabbit Polyclonal Antibody

0.1 mg
Catalog # 10068-506
Supplier:   Prosci
Fournisseur #:  79-227
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Catalog # 10068-506
Supplier:  Prosci
Anti-EIF2AK2 Rabbit Polyclonal Antibody
Anti-EIF2AK2 Rabbit Polyclonal Antibody

0.1 mg

Price
$934.17
Log in pour la disponibilité
Each (100µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Following activation by double-stranded RNA in the presence of ATP, the kinase becomes autophosphorylated and can catalyze the phosphorylation of the translation initiation factor EIF2S1, which leads to an inhibition of the initiation of protein synthesis. Double-stranded RNA is generated during the course of a viral infection.

Western Blot: 1:500~1:1000, Immunohistochemistry: 1:50~1:100, Immunofluorescence: 1:100~1:200

Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Spécifications
  • Taille:
    0.1 mg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Modification:
    Thr 446
  • Purification:
    Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
  • Antibody type:
    Primary
  • Form:
    Liquid
  • ImmunoFluorescence:
    Yes
  • Storage temperature:
    Store antibody at –20 °C for up to one year.
  • Host:
    Rabbit
  • Concentration:
    1 mg/mL
  • Molecular weight:
    68 kDa
  • Shipping temperature:
    4 °C
  • Antigen name:
    eukaryotic translation initiation factor 2-alpha kinase 2
  • Cat. No.:
    10068-506
  • Reactivity:
    Human
  • Gene ID:
    P19525
  • Antigen synonyms:
    EIF2AK2
  • Immunogen:
    PKR (Phospho-Thr446) antibody was raised against a peptide sequence around phosphorylation site of threonine 446 (K-R-T (p) -R-S) derived from Human PKR.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    EIF2AK2
  • Storage buffer:
    Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.