Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Anti-EIF4E3 Rabbit Polyclonal Antibody | Avantor

Anti-EIF4E3 Rabbit Polyclonal Antibody

100 ul
Catalog # 10108-450
Supplier:   Prosci
Fournisseur #:  29-517
Anti-EIF4E3 Rabbit Polyclonal Antibody
Catalog # 10108-450
Supplier:  Prosci
Anti-EIF4E3 Rabbit Polyclonal Antibody
Anti-EIF4E3 Rabbit Polyclonal Antibody

100 ul

Price
$1,250.37
Log in pour la disponibilité
Each (50µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.

EIF4E3 antibody can be used for detection of EIF4E3 by ELISA at 1:62500. EIF4E3 antibody can be used for detection of EIF4E3 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF4E3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Spécifications
  • Taille:
    100 ul
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Antibody is purified by peptide affinity chromatography method.
  • Antibody type:
    Primary
  • Form:
    Liquid
  • Storage temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store EIF4E3 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Host:
    Rabbit
  • Molecular weight:
    24 kDa
  • Shipping temperature:
    4 °C
  • Antigen name:
    eukaryotic translation initiation factor 4E family member 3
  • Cat. No.:
    10108-450
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    Q8N5X7
  • Antigen synonyms:
    EIF4E3
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human EIF4E3.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    EIF4E3
  • Storage buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.