Anti-CCBL2 Rabbit Polyclonal Antibody
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
RP11-82K18.3 is an aminotransferase that transaminates kynurenine to form kynurenic acid. Kynurenic acid is a metabolite of tryptophan.
RP11-82K18.3 antibody can be used for detection of RP11-82K18.3 by ELISA at 1:312500. RP11-82K18.3 antibody can be used for detection of RP11-82K18.3 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CCBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Spécifications
- Taille:100 ul
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antibody is purified by protein A chromatography method.
- Antibody type:Primary
- Form:Liquid
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store RP11-82K18.3 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Host:Rabbit
- Molecular weight:46 kDa, 51 kDa
- Shipping temperature:4 °C
- Antigen name:cysteine conjugate-beta lyase 2
- Cat. No.:10107-828
- Reactivity:Human
- Gene ID:Q6YP21
- Antigen synonyms:CCBL2
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human RP11-82K18.3.
- Conjugation:Unconjugated
- Antigen symbol:CCBL2
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



