pH Electrode Solutions, WTW
À propos de cet article
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
2 Options Available Below
Product Details & Documents
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 109648
- Supplier Number: 109705
- Saturated KCl, 125 mLDiscontinued-Supplier Number: -