Buffers, Ready to Use, WTW
Buffers, Ready to Use, WTW
Supplier:   WTW
Buffers, Ready to Use, WTW
Supplier:  WTW
Certificates
Buffers, Ready to Use, WTW
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Traceable to NIST
  • Reference temperature at 25 °C
  • 1 litre container with a separate compartment for the sample
Product Family Options
Informations sur le produit
  • Taille
    pH
    Availability
    Price
  • 1 L
    7.00
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (1l)
    Each (1l) - $229.41
    $229.41
    Each (1l)
       $229.41
    Supplier Number: 108702
  • 1 L
    4.01
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $219.25
    $219.25
    Each
       $219.25
    Supplier Number: 108700
  • 1 L
    10.01
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $238.88
    $238.88
    Each
       $238.88
    Supplier Number: 108703