Cables for Electrodes, WTW
Cable with S7/DIN plug, 1 m (3.3 ft.)
Catalog # 80085-308
Supplier: WTW
Fournisseur #: 108110
Jump to:
Product Details & Documents
Spécifications
- Désignation:Cable with S7/DIN plug
- Longueur:1 m (3.3 ft.)