CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific | Avantor
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
Supplier:  Globe Scientific
Certificates
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
6 Options Available Below
Product Details & Documents
Innovation to protect precious samples.
  • Non-pyrogenic, non-cytotoxic, non-mutagenic, heavy metals free, BSE/TSE free
  • RNase, DNase, ATP, and Human DNA free
  • Tamper evident packaging
  • Autoclavable
The best and most leak-resistant cryogenic vials available for the long-term storage of your precious samples at temperatures as low as –196 °C.
The vials feature an innovative screw cap that is co-molded with a layer of thermoplastic elastomer (TPE). This special process provides the leak-proof benefits of an O-ring, while eliminating the risk of contamination and sample loss associated with the use of separate O-rings.
Each vial is printed with a unique barcode that may be used for automated data collection, accurate sample inventory or to conceal the sample's identity.
Packaging: Packaged in tamper-evident bags of 50.
Caution: Not for use in the liquid phase of liquid nitrogen.
Product Family Options
Informations sur le produit
  • Capacité
    Bottom Style
    Avec bouchon
    Sterility
    Availability
    Price
  • 1.00 ml
    Conical bottom, self-standing
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $536.50
    $536.50
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $536.50
    Supplier Number: 3001
  • 2.00 ml
    Round bottom, self-standing
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $502.48
    $502.48
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $502.48
    Supplier Number: 3002
  • 4.00 ml
    Round bottom
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $617.89
    $617.89
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $617.89
    Supplier Number: 3004
  • 4.00 ml
    Round bottom, self-standing
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $622.45
    $622.45
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $622.45
    Supplier Number: 3005
  • 5.00 ml
    Round bottom
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $675.10
    $675.10
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $675.10
    Supplier Number: 3006
  • 5.00 ml
    Round bottom, self-standing
    Oui
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $681.34
    $681.34
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $681.34
    Supplier Number: 3008
  • Round bottom
    Discontinued
    Supplier Number: -