Shake Flasks, Wheaton
Shake Flasks, Wheaton
Shake Flasks, Wheaton
Certificates
Shake Flasks, Wheaton
7 Options Available Below
Product Details & Documents
Wheaton Erlenmeyer Shake Flasks are ideal for all suspension cell cultures, including baculovirus, microbial and algae cultures, as well as media preparation, storage, and all related applications.
  • Gamma irradiated
  • Wrapped in an easy-tear, sterile bag
  • Fit all standard shaking incubator systems and can be placed on any platform
Lexan polycarbonate resin is an amorphous engineering thermoplastic, characterized by outstanding mechanical, optical, electrical, and thermal properties. The Lexan resins utilized by Wheaton environmentally conform to flame retardant standards, are scratch resistant, with a toughness and heat resistance that ensures biocompatible and optical quality. Wheaton products made with the Lexan polycarbonate resin are compliant with stringent FDA and USP requirements. Based on these properties and their inert nature, the media bottles and shaker flasks are ideal packaging vessels. All products are manufactured in a Class 10K cleanroom.
In addition, Wheaton offers shaker flask clamps. The polycarbonate products are supplied gamma irradiated sterilized (SAL> 10¯⁶). All products are packaged and produced in a cleanroom controlled environment and do not need to be rinsed prior to first use. Consequently, all Lexan polycarbonate containers are a safer alternative to glass. Polycarbonate Erlenmeyer flasks can withstand repeated autoclaving and are optimal for all microbial culture. Wheaton Erlenmeyer flasks are certified pyrogen-, DNase-, and RNase-free. They are packaged particulate-free and wrapped in a sterile easy-tear bag.
Product Family Options
Flat base
Product Information
  • Capacité
    Epaisseur
    Availability
    Price
  • 125 ml
    38 - 430
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $400.35
    $400.35
    New Customer Promotional Price
    Case of 24
       $400.35
    Supplier Number: WPFPC0125S
  • 250 ml
    38 - 430
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $235.45
    $235.45
    New Customer Promotional Price
    Case of 12
       $235.45
    Supplier Number: WPFPC0250S
  • 500 ml
    38 - 430
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $317.05
    $317.05
    New Customer Promotional Price
    Case of 12
       $317.05
    Supplier Number: WPFPC0500S
  • 1000 ml
    53B
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $234.60
    $234.60
    New Customer Promotional Price
    Case of 6
       $234.60
    Supplier Number: WPFPC1000S
  • 2000 ml
    53B
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $280.50
    $280.50
    New Customer Promotional Price
    Case of 6
       $280.50
    Supplier Number: WPFPC2000S
Baffled
Product Information
  • Capacité
    Epaisseur
    Availability
    Price
  • 125 ml
    38 - 430
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $400.35
    $400.35
    New Customer Promotional Price
    Case of 24
       $400.35
    Supplier Number: WPFBC0125S
  • 250 ml
    38 - 430
    Log in for availability
    Log in for availability
    Case of 12
    Case of 12 - $235.45
    $235.45
    New Customer Promotional Price
    Case of 12
       $235.45
    Supplier Number: WPFBC0250S
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Polypropylene Clamp for 2000 mL Shake Flask, 169.1 dia. X 89.57 mm
    Polypropylene Clamp for 2000 mL Shake Flask, 169.1 dia. X 89.57 mm
    10112-158
    Log in for availability
    Case of 1 - $204.75
Product Information
  • Catalog #
    Availability
    Price
  • Polypropylene Clamp for 500 mL Shake Flask, 111.6 dia. X 63.14 mm
    Polypropylene Clamp for 500 mL Shake Flask, 111.6 dia. X 63.14 mm
    10112-162
    Log in for availability
    Case of 1 - $153.79
Product Information
  • Catalog #
    Availability
    Price
  • Polypropylene Clamp for 1000 mL Shake Flask, 143.9 dia. X 74.5 mm
    Polypropylene Clamp for 1000 mL Shake Flask, 143.9 dia. X 74.5 mm
    10112-160
    Log in for availability
    Case of 1 - $182.91
Product Information
  • Catalog #
    Availability
    Price
  • Polypropylene Clamp for 250 mL Shake Flask, 90.4 dia. X 57.63 mm
    Polypropylene Clamp for 250 mL Shake Flask, 90.4 dia. X 57.63 mm
    10112-164
    Log in for availability
    Case of 1 - $131.95
Product Information
  • Catalog #
    Availability
    Price
  • Polypropylene Clamp for 125 mL Shake Flask, 74.72 dia. X 51.17 mm
    Polypropylene Clamp for 125 mL Shake Flask, 74.72 dia. X 51.17 mm
    10112-166
    Log in for availability
    Case of 1 - $117.39