Burn-Up Box, Dynalon
Catalogue # 56765-965
Fournisseur: Dynalon
Spécifications
- Dimensions:12×12×20" (L×W×H); Ø 7"
- Cat. no.:56765-965
- Supplier No.:797065
Spécifications
À propos de cet article
Burn-Up Box features colorful graphics to allow for quick identification
Boxes fold for economical shipping and storage. Closure assures safe, hygienic disposal. Supplied with tough 2 millimeter polyethylene liner.