About this item
CTC PAL autosamplers are one of the most popular sampling devices on the market today
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. S-Line syringes deliver great performance in everyday use for HPLC injections at an exceptional value. The Gastight® S-Line syringe is an ideal choice for HPLC sample injections. It incorporates a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit in the glass barrel, the tip wipes the interior of the syringe barrel free of sample, minimizing sample carryover. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Customers rely on their ability to sample from many different vial and well types and on their ultimate platform flexibility. The syringe is the heart of every injection that the HPLC PAL system performs. The latest generation of Hamilton autosampler syringes is recommended by CTC for use in the popular HPLC PAL systems: PAL HTC = xt, PAL HTS = xt, PAL HTX = xt.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 89498-976
- 89498-978
- 89498-980
- 89498-982
- 89498-972
- 89498-974
- Type:
- Gastight
- Gastight
- Gastight
- Gastight
- Gastight
- Gastight
- Series:
- 1700
- 1700
- 1700
- 1700
- 1700
- 1700
- Autoclavable:
- No
- No
- No
- No
- No
- No
- Ext.Ø:
- 0.711 mm (Needle)
- 0.711 mm (Needle)
- 0.711 mm (Needle)
- 0.711 mm (Needle)
- 0.711 mm (Needle)
- 0.711 mm (Needle)
- Needle length:
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- 51 mm
- Point style:
- 3
- 3
- 3
- 3
- 3
- 3
- Termination:
- Cemented Needle (N)
- Cemented Needle (N)
- Cemented Needle (N)
- Cemented Needle (N)
- Cemented Needle (N)
- Cemented Needle (N)