About this item
The PHCbi Twin Guard® Series satisfies an industry demand for safe, long-term storage for the most high-valued materials.
- Two refrigeration systems
- Ideal for storing sensitive specimens
- EcoMode functionality
- Five-year manufacturer's parts and labor warranty
Two independent refrigeration systems, combined with an optional liquid CO₂ back-up system, offer a circle of protection unmatched in the marketplace
The two TwinGuard® Series' compressors are completely separated from one another with no reliance on one another to operate. In the unlikely event of a failure, the other compressor continues to function at ultra-low temperatures. TwinGuard® Series freezers also work to minimize energy costs through a unique EcoMode function while providing optimum chamber uniformity of ±5 °C over a wide operating range from –50 to –86 °C. TwinGuard® freezers help lower operational costs with a small foot print and reduced energy usage.
Developed for use with conventional inventory racks and boxes, the TwinGuard® Series is ideal for storage of sensitive stem cells, embryos, cell lines, and other rare specimens.
Specifications
- Catalog No:
- CA76019-348
- CA76200-148
- CA76019-346
- Model:
- MDF-DU702VXCPA
- MDF-DU302VX-PA
- MDF-DU502VXCPA
- Cat. No.:
- 76019-348
- 76200-148
- 76019-346
- Insulation:
- Patented VIP® PLUS Composite Wall, Energy Savings
- Patented VIP® PLUS Composite Wall, Energy Savings
- Patented VIP® PLUS Composite Wall, Energy Savings
- Electrical:
- 208/230 V, NEMA 6-15
- 208/230 V, NEMA 6-15
- 208/230 V, NEMA 6-15
- Shelves/drawers:
- 3 adjustable stainless steel shelves, 4 compartments
- 3 adjustable stainless steel shelves, 4 compartments
- 3 adjustable stainless steel shelves, 4 compartments
- Weight:
- 383 kg (844 lbs.)
- 225 kg (496 lbs.)
- 328 kg (723 lbs.)