Brooms, Stiff, Remco Products
Supplier: Remco Products
Certificates
About this item
This heavy duty broom has long, thick bristles, which makes it ideal for sweeping heavy debris
This brush is also suitable for use in wet and external areas. Block material is polypropylene; bristle material is PET.
Certifications: FDA Compliant Raw Material.
Frequently Bought Together