ContactUsLinkComponent
Beakers, PTFE, Dynalon
  13914-541
 :  Dynalon
 : 312044
undefined
Beakers, PTFE, Dynalon
  13914-541
 :  Dynalon
 :  312284
 : 312044
Select an Option
43 mm
Select an Option
50 ml (1.7 oz.)
Select an Option
60 mm (2.4")
Select an Option
13914-541
Select an Option
0.07875 lbs.
Select an Option
312044
Isostatically molded beakers are manufactured from virgin PTFE and machine finished to give a smooth surface for easy cleaning and contamination elimination.
  • With pouring spout
  • Working temperature range is from cryogenic to 260°C
Virtually immune to chemical attack, beakers are only attacked by some fluorine containing compounds at high temperatures, and molten alkalis metals.
Frequently Bought Together