Stir Bar, Triangular, PTFE, Dynalon
Supplier: Dynalon
Jump to:
À propos de cet article
Triangular shape provides the best possible spinning action and most efficient mixing of all stir bars
Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.
2 Options Available Below
Product Details & Documents
Triangular shape provides the best possible spinning action and most efficient mixing of all stir bars
Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.