Anti-IgG Rabbit Polyclonal Antibody
Catalogue # RL-602-4102
Fournisseur: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Cat
- Western blot:Yes
- Size:2 mg
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Concentration:1.9 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Cat IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:RL-602-4102
- Supplier No.:602-4102
Spécifications
À propos de cet article
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Cat