Anti-PAP3 Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemical
À propos de cet article
Recommended Dilutions: ELISA: 1:4000 - 1:40000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000
Type: Primary
Antigen: PAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Potato
2 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Recommended Dilutions: ELISA: 1:4000 - 1:40000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000
Type: Primary
Antigen: PAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Potato
Specifications for Product Family
Spécifications
- Catalog No:
- CARL2004140100
- CARL2004140S
- Antigen symbol:
- PAP3
- PAP3
- Antigen name:
- Purple Acid phosphatase 3
- Purple Acid phosphatase 3
- Antigen synonyms:
- Cytoplasmic phosphotyrosyl protein phosphatase|Acid phosphatase 1 soluble|HAAP|Acid phosphatase of erythrocyte|Adipocyte acid phosphatase
- Cytoplasmic phosphotyrosyl protein phosphatase|Acid phosphatase 1 soluble|HAAP|Acid phosphatase of erythrocyte|Adipocyte acid phosphatase
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Potato
- Potato
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Acid Phosphatase [Potato]
- Acid Phosphatase [Potato]
- Purification:
- purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Concentration:
- by UV absorbance at 280 nm
- by UV absorbance at 280 nm
- Cross adsorption:
- No
- No
- Storage buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Dry Ice
Spécifications
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- Supplier Number: 200-4140-0100L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.CARL2004140100Antigen symbolPAP3Antigen namePurple Acid phosphatase 3Antigen synonymsCytoplasmic phosphotyrosyl protein phosphatase|Acid phosphatase 1 soluble|HAAP|Acid phosphatase of erythrocyte|Adipocyte acid phosphataseAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityPotatoHostRabbitWestern blotYesELISAYesImmunoChemistryYesImmunogenAcid Phosphatase [Potato]Purificationpurified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the bufferConcentrationby UV absorbance at 280 nmCross adsorptionNoStorage buffer0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium AzideStorage temperature–20 °CShipping temperatureAmbient - Supplier Number: 200-4140SL'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.CARL2004140SAntigen symbolPAP3Antigen namePurple Acid phosphatase 3Antigen synonymsCytoplasmic phosphotyrosyl protein phosphatase|Acid phosphatase 1 soluble|HAAP|Acid phosphatase of erythrocyte|Adipocyte acid phosphataseAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityPotatoHostRabbitWestern blotYesELISAYesImmunoChemistryYesImmunogenAcid Phosphatase [Potato]Purificationpurified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the bufferConcentrationby UV absorbance at 280 nmCross adsorptionNoStorage buffer0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium AzideStorage temperature–20 °CShipping temperatureDry Ice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



