Round Dipping Baskets, Dynalon
Round dipping baskets, fully perforated and with swing handle, are fabricated of either 1/8” thick HDPE or PP.
HDE takes 140 °F on a continuous basis or more for shorter periods (up to 160°F). Polypropylene fabricated dipping baskets withstands 180°F on a continuous basis or up to 200 °F for shorter periods. Both are supplied with sturdy swinging handle—a unique feature allowing easy storage and loading. All have perforated bottoms. Excellent chemical resistance.
Ordering information: Additional sizes and materials are available upon request.
Round dipping baskets, fully perforated and with swing handle, are fabricated of either 1/8” thick HDPE or PP.
HDE takes 140 °F on a continuous basis or more for shorter periods (up to 160°F). Polypropylene fabricated dipping baskets withstands 180°F on a continuous basis or up to 200 °F for shorter periods. Both are supplied with sturdy swinging handle—a unique feature allowing easy storage and loading. All have perforated bottoms. Excellent chemical resistance.
Ordering information: Additional sizes and materials are available upon request.
Product Family Options
- DésignationMatièreDimensionsWeight
- Round Dipping Baskets, Perforations 1/8" Dia. on 21/64" CenterPP6×6" (OD×H)0.85 lbs.-: 146604
- Round Dipping Baskets, Perforations 1/8" Dia. on 21/64" CenterPP12×12" (OD×H)3.0 lbs.-: 146644
- Round Dipping Baskets, Perforations 1/8" Dia. on 21/64" CenterHDPE12×12" (OD×H)2.65 lbs.Each$652.19: 146594