ContactUsLinkComponent
Plastic Bag Ties, 4000 Foot Spool, Dynalon
Plastic Bag Ties, 4000 Foot Spool, Dynalon
Catalog # 71070-260
Supplier:  Dynalon
undefined
Plastic Bag Ties, 4000 Foot Spool, Dynalon
Catalog # 71070-260
Supplier:  Dynalon
Supplier Number:  626814
Specifications
  • Cat. no.:
    71070-260
  • Weight:
    4.4 lbs.
  • Supplier No.:
    626814
Specifications
About this item
Plastic coated bag ties are extruded of bright red LDPE bonded to an annealed steel wire.
Can be cut to any size.