Cuvette Cap, Square, Dynalon
¼", ½", ½", 0.00062 lbs
Catalog # 30620-274
Supplier: Dynalon
Supplier #: 402364
Jump to:
Product Details & Documents
Cuvette caps, polyethylene dust covers fit most standard cuvettes.
Flexible fins on cap allow for tight fit that will not damage the cuvette. Caps are 19 mm in depth.
Specifications
- Largeur:½"
- Hauteur:¼"
- Longueur:½"
- Weight:0.00062 lbs