81-Well Microtube Storage Boxes
Made of translucent, vibrantly colored polypropylene which allows for visibility inside the box and color coding identification. Tube capacity is indicated on the outside the box within a distinctive wave design. Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area.
- Easy open and secure lid fitted with friction fit design
- Stackable, saving valuable space in the lab
- Can be used in temperatures as low as –80 °C
- Autoclavable
Featuring molded grid lines on the lid, imprinted coordinates on the base, and a molded reference point on the lid.
Holds 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials.
Frequently Bought Together