MBP Molecular Devices® -Style 384-Well Tips, Thermo Scientific
Catalog # 89108-097
Supplier: Thermo Scientific
Specifications
- Volume:30 µl
- Cat. no.:89108-097
- Tip Style:Clear, Pre-sterilized
- Supplier No.:931-261
Specifications
About this item
MBP leads the way in FLIPR style tips with our SoftFit technology.
- Thin wall design that creates a better seal, reduces the loading force required
- Improves %CVs, and reduces wear and tear on the machine
ART is a registered trademark of MBP and is protected by U.S. Patent # 5,156,811. SoftFit is a trademark of Molecular Bio-Product. Molecular Devices and FLIPR are registered trademarks of Molecular Devices Corp.