MBP MicroPoint Pipette Tip, 300 µl, Thermo Scientific
Catalog # 53225-706
Supplier: Thermo Scientific
Specifications
- Volume:300 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Cat. no.:53225-706
- Sterile:+
- Tip Style:MicroPoint Pipette Tip, Racked
- Supplier No.:3571
Specifications
About this item
Perform low-volume sample work with Thermo Scientific ART non-filtered pipette tips with Micropoint tip design, which feature a tip profile, nearly 50% smaller than standard blunt-end tips and significantly more narrow than beveled-end tips.
Crystal clear tips allow transparent samples to be seen easily.