VWR® 4 Well PCR Reaction Strips and Caps
Fournisseur: VWR International
Certificats
À propos de cet article
These 4 well ultra clear reaction strips fit the Qiagen® Rotorgene®.
- Polypropylene fabrication
RNase/DNase-free.
Delivery: They include 250 4-well strips and 250 separate strip caps per package.
Spécifications
- Catalog No:
- 76318-806
- 89218-300
- Environmentally Preferable:
- Sustainable Packaging, Low Manufacturing Impact