VWR® Isolator Tool Cover
Catalog # 10029-772
Supplier: VWR International
Small Business Enterprise
Specifications
- Description:Microfiber cover, blue
- Length:9" (23 cm)
- Material:Blue microfiber, 80% polyester/20% polyamide
- Cat. no.:10029-772
- Details:Elastic band attachment
Specifications
About this item
An elasticized microfiber slip cover for use with the QDIT/QPIT series mop adapters.
- Microfiber
- Lot numbers printed on each bag and each case
Designed for isolators, bio-hazard and containment hoods.
Certifications: ISO 9001:2008 Facility.
Packaging: Each mop head single bagged and packaged 50 mops per poly lined case.